3,4,5-Trimethoxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethylgallic Acid

13993-0050 13993-1000 13993-5000
TS13993-0050EA 21 USD
TS13993-0050 TS13993-1000 TS13993-5000
3,4,5-Trimethoxybenzoic acid 99%
3,4,5-Trimethoxybenzoic acid
Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
Boiling Pt: 225…227 °C (10 mmHg)
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00002501
CAS Number: 118-41-2
EINECS: 204-248-2

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 172 °C
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR