3,4,5-Trimethoxybenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

13991-0250 13991-1000 13991-5000
TS13991-0250EA 38 USD
TS13991-0250 TS13991-1000 TS13991-5000
3,4,5-Trimethoxybenzaldehyde 99%
3,4,5-Trimethoxybenzaldehyde
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 163…165 °C (10 mmHg)
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00003364
CAS Number: 86-81-7
EINECS: 201-701-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to cream Powder or flakes
Infrared spectrum Conforms
Melting point 73 to 76 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR