Triethylene glycol 99%

Supplier: Thermo Scientific Chemicals

13959-0010 13959-0025 13959-0100
TS13959-0010EA 73 USD
TS13959-0010 TS13959-0025 TS13959-0100
Triethylene glycol 99%
Triethylene glycol
Formula: C₆H₁₄O₄
Boiling Pt: 285 °C (760 mmHg)
Melting Pt: –7 °C
Density: 1.12 g/cm³
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002880
CAS Number: 112-27-6
EINECS: 203-953-2
Merck Index: 12,09802

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4545 to 1.4565 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR