2,4,6-Trichlorophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

13948-1000 13948-5000
TS13948-1000EA 44 USD
TS13948-1000 TS13948-5000
2,4,6-Trichlorophenol 98%
2,4,6-Trichlorophenol
Formula: C₆H₃Cl₃O
Boiling Pt: 246 °C (760 mmHg)
Melting Pt: 64…68 °C
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002172
CAS Number: 88-06-2
EINECS: 201-795-9
UN: 2020
ADR: 6.1,III
Merck Index: 12,09773

Order Now

Specification Test Results

Appearance Light beige to light brown Crystalline powder or mass and/or chunks
Infrared spectrum Conforms
Melting point 64 to 68 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR