p-Tolylhydrazinium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylphenylhydrazinium chloride, 4-Tolylhydrazine hydrochloride, p-Tolylhydrazine hydrochloride, 1-(4-Methylphenyl)hydrazine hydrochloride, 4-Methylphenylhydrazine hydrochloride, 1-(4-Tolyl)hydrazine hydrochloride

13916-0250 13916-1000
TS13916-0250EA 122 USD
TS13916-0250 TS13916-1000
p-Tolylhydrazinium chloride 98%
p-Tolylhydrazinium chloride
Formula: C₇H₁₁ClN₂
Melting Pt: 241 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012940
CAS Number: 637-60-5
EINECS: 211-295-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to beige to light brown Powder or flakes
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR