m-Toluic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methylbenzoic acid

13905-0010 13905-0050 13905-1000
TS13905-0010EA 124 USD
TS13905-0010 TS13905-0050 TS13905-1000
m-Toluic acid 99%
m-Toluic acid
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 108…112 °C
Density: 1.054 g/cm³
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002523
CAS Number: 99-04-7
EINECS: 202-723-9
Merck Index: 12,09673

Order Now

Specification Test Results

Appearance Light yellow to light beige Crystalline solid
Infrared spectrum Conforms
Melting point 108 to 112 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR