3-Tolualdehyde 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
13899-0250
13899-1000
TS13899-0250EA
83
USD
TS13899-0250
TS13899-1000
3-Tolualdehyde 98% stabilized
3-Tolualdehyde
Formula:
C₈H₈O Boiling Pt: 199 °C (1013 hPa) Density: 1.019 g/cm³ Flash Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003374 CAS Number: 620-23-5 EINECS: 210-632-0 |
Specification Test Results
Appearance | Clear yellow to brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5400 to 1.5420 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | 0.1 % hydroquinone |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...