2-Acetyl-6-methoxynaphthalene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6'-Methoxy-2'-acetonaphthone

13478-0010 13478-0100
TS13478-0010EA 20 USD
TS13478-0010 TS13478-0100
2-Acetyl-6-methoxynaphthalene 98%
2-Acetyl-6-methoxynaphthalene
Formula: C₁₃H₁₂O₂
Melting Pt: 108…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00021643
CAS Number: 3900-45-6

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 106 to 109 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR