α,2,4-Trichlorotoluene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,4-Dichlorobenzyl chloride, 1-(Chloromethyl)-2,4-dichlorobenzene, 1,3-Dichloro-4-(chloromethyl)benzene, 2,4-Dichloro-1-(chloromethyl)benzene

13439-0050 13439-1000 13439-5000
TS13439-0050EA 29 USD
TS13439-0050 TS13439-1000 TS13439-5000
α,2,4-Trichlorotoluene 99%
α,2,4-Trichlorotoluene
Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: –2.7 °C
Density: 1.4 g/cm³
Flash Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00000895
CAS Number: 94-99-5
EINECS: 202-381-0
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5750 to 1.5770 (20°C, 589 nm)
GC ≥98.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation +7° to +9° (20°C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR