4-Iodotoluene 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Iodo-4-methylbenzene

13394-0100 13394-0500 13394-1000
TS13394-0100EA 42 USD
TS13394-0100 TS13394-0500 TS13394-1000
4-Iodotoluene 98%
4-Iodotoluene
Formula: C₇H₇I
MW: 218.04 g/mol
Boiling Pt: 211.5 °C (1013 hPa)
Melting Pt: 33…36 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001059
CAS Number: 624-31-7
EINECS: 210-841-7

Order Now

Specification Test Results

Appearance White to light yellow Low melting crystalline mass
Infrared spectrum Conforms
Melting point 33 to 36 °C
GC ≥97.5 %
Elemental Analysis 7.55 to 8.35 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR