Salicylhydrazide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxybenzoic acid hydrazide, Salicylohydrazide, 2-Hydroxybenzene-1-carbohydrazide, Salicylic acid hydrazide, 2-Hydroxybenzohydrazide, 2-Hydroxybenzhydrazide, 2-Hydroxybenzoic hydrazide
13261-0250
13261-1000
TS13261-0250EA
69
USD
TS13261-0250
TS13261-1000
Salicylhydrazide 98+%
Salicylhydrazide
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 147…150 °C |
MDL Number:
MFCD00007599 CAS Number: 936-02-7 EINECS: 213-311-3 |
Specification Test Results
Appearance | White to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 146 to 151 °C |
Titration with HClO4 | ≥98.0 % |
Specific optical rotation | -14° to -18° (20 °C, 589 nm) (c=1, ethanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...