Resorcinol 98%

Supplier: Thermo Scientific Chemicals
Warning

13229-0010 13229-0050 13229-0500 13229-2500
TS13229-0010EA 128 USD
TS13229-0010 TS13229-0050 TS13229-0500 TS13229-2500
Resorcinol 98%
Resorcinol
Formula: C₆H₆O₂
Boiling Pt: 281 °C (760 mmHg)
Melting Pt: 109…111 °C
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00002269
CAS Number: 108-46-3
EINECS: 203-585-2
UN: 2876
ADR: 6.1,III
Merck Index: 12,08323
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69fa2900537bdb42ccfd55e4318288eb881a61a0&vid=e62c617319ff4711a7ff00e69774ad92cfd75d09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to yellow to beige to pink Crystals or granules or flakes
Infrared spectrum Conforms
Melting point 108 to 112 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR