Phthalic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pathalic acid

13107-0010 13107-0025 13107-2500
TS13107-0010EA 133 USD
TS13107-0010 TS13107-0025 TS13107-2500
Phthalic acid 99%
Phthalic acid
Formula: C₈H₆O₄
MW: 166.13 g/mol
Melting Pt: 200…205 °C (dec.)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00002467
CAS Number: 88-99-3
EINECS: 201-873-2
Merck Index: 12,07527

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 200 to 205 °C (Dec)
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR