1-Phenyl-2-urea 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenylurea, 1-Phenylurea, N-Phenylurea

13102-0050 13102-1000 13102-2500
TS13102-0050EA 21 USD
TS13102-0050 TS13102-1000 TS13102-2500
1-Phenyl-2-urea 97%
1-Phenyl-2-urea
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 143…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007944
CAS Number: 64-10-8
EINECS: 200-576-5

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 143 to 149 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR