Diphenyl sulfone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenyl sulphone, sulphonyldibenzene

13096-0010 13096-0050 13096-2500
TS13096-0010EA 142 USD
TS13096-0010 TS13096-0050 TS13096-2500
Diphenyl sulfone 97%
Diphenyl sulphone
Formula: C₁₂H₁₀O₂S
MW: 218.28 g/mol
Melting Pt: 125…129 °C
MDL Number: MFCD00007548
CAS Number: 127-63-9
EINECS: 204-853-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 129 °C
GC ≥96.0 %
Stabilizer Approx. 50 ppm BHT
Peroxide value ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR