2-Phenyl-2-propanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α,α-Dimethylbenzyl alcohol

13083-0050 13083-0250 13083-1000
TS13083-0050EA 52 USD
TS13083-0050 TS13083-0250 TS13083-1000
2-Phenyl-2-propanol 99%
2-Phenyl-2-propanol
Formula: C₆H₅C(OH)(CH₃)₂
MW: 136.19 g/mol
Boiling Pt: 88…90 °C (11 mmHg)
Melting Pt: 32…34 °C
Density: 0.973 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00004456
CAS Number: 617-94-7
EINECS: 210-539-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f8edbb976395240a99a4f52addfbfe32e8c926f&vid=5b4ae0c8b87b317b3d1326768c6ff8bdaaab37f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to white to yellow Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5190 to 1.5220 (20°C, 589 nm)
GC ≥98.5 %
UV at 335 nm A: ≤1.0
Color scale ≤15 APHA
Residue after evaporation ≤0.001 %
Acidity/alkalinity ≤0.0005 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR