Biphenyl-2-ol 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenylphenol, 2-Hydroxydiphenyl, 2-Hydroxybiphenyl

13076-0025 13076-0050 13076-5000
TS13076-0025EA 265 USD
TS13076-0025 TS13076-0050 TS13076-5000
Biphenyl-2-ol 99+%
Biphenyl-2-ol
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 280…286 °C (1013 hPa)
Melting Pt: >56 °C
Density: 1.21 g/cm³
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD00002208
CAS Number: 90-43-7
EINECS: 201-993-5
UN: 3077
ADR: 9,III
Merck Index: 12,07458

Order Now

Specification Test Results

Appearance White to off-white Crystalline flakes
Infrared spectrum Conforms
Melting point 56 to 59 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR