Diphenyl ether 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diphenyl oxide, Phenyl ether, Phenoxybenzene

13060-0010 13060-0025 13060-0100 13060-1000 13060-5000
TS13060-0010EA 63 USD
TS13060-0010 TS13060-0025 TS13060-0100 TS13060-1000 TS13060-5000
Diphenyl ether 99%
Diphenyl ether
Formula: C₁₂H₁₀O
Boiling Pt: 259 °C (760 mmHg)
Melting Pt: 26…30 °C
Density: 1.073 g/cm³
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00003034
CAS Number: 101-84-8
EINECS: 202-981-2
UN: 3077
ADR: 9,III
Merck Index: 12,07442

Order Now

Specification Test Results

Appearance Clear light yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5795 to 1.5815 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR