4-Phenoxyaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Amino-4-phenoxybenzol

13023-0050 13023-1000 13023-5000
TS13023-0050EA 20 USD
TS13023-0050 TS13023-1000 TS13023-5000
4-Phenoxyaniline 97%
4-Phenoxyaniline
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 140 °C (3hPa)
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00007862
CAS Number: 139-59-3
EINECS: 205-367-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Brown to green-brown Powder or flakes
Infrared spectrum Conforms
Melting point 82 to 86 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR