1,5-Pentanediol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Pentane-1,5-diol, 1,5-Dihydroxy pentane

12995-0010 12995-0050 12995-1000 12995-5000
TS12995-0010EA 116 USD
TS12995-0010 TS12995-0050 TS12995-1000 TS12995-5000
1,5-Pentanediol 98%
1,5-Pentanediol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 242 °C (760 mmHg)
Melting Pt: –18 °C
Density: 0.994 g/cm³
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002978
CAS Number: 111-29-5
EINECS: 203-854-4
Merck Index: 12,07256

Order Now

Specification Test Results

Appearance Clear colorless Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20°C, 589 nm)
GC ≥97.5 %
Elemental Analysis 14.96 to 16.17 % N
Residue after evaporation ≤0.01 %
Peroxides ≤50 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR