1,9-Nonanediol 99%

Supplier: Thermo Scientific Chemicals

12912-0250 12912-1000
TS12912-0250EA 45 USD
TS12912-0250 TS12912-1000
1,9-Nonanediol 99%
1,9-Nonanediol
Formula: C₉H₂₀O₂
Boiling Pt: 177 °C (15 mmHg)
Melting Pt: 44…48 °C
Density: 0.918 g/cm³
Flash Pt: 162 °C
Storage Temperature: Ambient
MDL Number: MFCD00002991
CAS Number: 3937-56-2
EINECS: 223-517-5

Order Now

Specification Test Results

Appearance White Fused crystalline solid
Infrared spectrum Conforms
Melting point 44 to 48 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR