3-Nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-Nitrotoluene

12904-0050 12904-5000
TS12904-0050EA 31 USD
TS12904-0050 TS12904-5000
3-Nitrotoluene 99%
3-Nitrotoluene
Formula: C₇H₇NO₂
Boiling Pt: 230…231 °C (760 mmHg)
Melting Pt: 15…16 °C
Density: 1.15 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007265
CAS Number: 99-08-1
EINECS: 202-728-6
UN: 1664
ADR: 6.1,II
Merck Index: 12,06748

Order Now

Specification Test Results

Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR