2-Nitrotoluene 99+%

Supplier: Thermo Scientific Chemicals
Danger

12903-0010 12903-0025
TS12903-0010EA 74 USD
TS12903-0010 TS12903-0025
2-Nitrotoluene 99+%
2-Nitrotoluene
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 222 °C (760 mmHg)
Melting Pt: –9 °C
Density: 1.16 g/cm³
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00007157
CAS Number: 88-72-2
EINECS: 201-853-3
UN: 1664
ADR: 6.1,II
Merck Index: 12,06748

Order Now

Specification Test Results

Appearance Clear yellow to yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.5445 to 1.5465 (20°C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR