α-Bromo-4-nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Nitrobenzyl bromide, 1-(Bromomethyl)-4-nitrobenzene, α-Bromo-p-nitrotoluene

12853-0250 12853-1000 12853-5000
TS12853-0250EA 46 USD
TS12853-0250 TS12853-1000 TS12853-5000
α-Bromo-4-nitrotoluene 99%
α-Bromo-4-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 98…100 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007373
CAS Number: 100-11-8
EINECS: 202-820-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 100 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR