4-Nitrobenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Nitrophenyl)methanol

12852-0250 12852-1000
TS12852-0250EA 42 USD
TS12852-0250 TS12852-1000
4-Nitrobenzyl alcohol 99%
4-Nitrobenzyl alcohol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 185 °C (12 mmHg)
Melting Pt: 92…95 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00007376
CAS Number: 619-73-8
EINECS: 210-611-6

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 95 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR