3-Nitrobenzaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

12841-1000 12841-5000
TS12841-1000EA 39 USD
TS12841-1000 TS12841-5000
3-Nitrobenzaldehyde 99%
3-Nitrobenzaldehyde
Formula: C₇H₅NO₃
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00007249
CAS Number: 99-61-6
EINECS: 202-772-6
UN: 3077
Merck Index: 12,06684

Order Now

Specification Test Results

Appearance Yellow to yellow-brown Granular powder
Infrared spectrum Conforms
Melting point 56 to 59 °C
Water ≤0.5 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR