2-Nitrobenzaldehyde 99+%

Supplier: Thermo Scientific Chemicals
Warning

12840-0250 12840-1000 12840-5000
TS12840-0250EA 68 USD
TS12840-0250 TS12840-1000 TS12840-5000
2-Nitrobenzaldehyde 99+%
2-Nitrobenzaldehyde
Formula: C₇H₅NO₃
Boiling Pt: 153 °C (23 mmHg)
Melting Pt: 41…44 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007132
CAS Number: 552-89-6
EINECS: 209-025-3
Merck Index: 12,06684

Order Now

Specification Test Results

Appearance Yellow Powder or needles and/or chunks
Infrared spectrum Conforms
Melting point 41 to 45 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR