1,8-Naphthalenedicarboxylic anhydride 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Naphthalene-1,8-dicarboxylic anhydride, Benzo[de]isochromene-1,3-dione, 1,8-Naphthalic anhydride

12814-0025 12814-1000 12814-5000
TS12814-0025EA 371 USD
TS12814-0025 TS12814-1000 TS12814-5000
1,8-Naphthalenedicarboxylic anhydride 97%
1,8-Naphthalenedicarboxylic anhydride
Formula: C₁₂H₆O₃
Melting Pt: >269 °C
Flash Pt: 272 °C
Storage Temperature: Ambient
MDL Number: MFCD00006925
CAS Number: 81-84-5
EINECS: 201-380-2

Order Now

Specification Test Results

Appearance Beige to light brown Powder
Infrared spectrum Conforms
Melting point ≥269 °C
Loss on drying ≤0.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR