α-Methylstyrene 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Phenylpropene, Isopropenylbenzene

12771-0010 12771-0025 12771-1000
TS12771-0010EA 50 USD
TS12771-0010 TS12771-0025 TS12771-1000
α-Methylstyrene 99% stabilized
α-Methylstyrene
Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 165…169 °C (760 mmHg)
Melting Pt: –23 °C
Density: 0.909 g/cm³
Flash Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008859
CAS Number: 98-83-9
EINECS: 202-705-0
UN: 2303
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5390 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer 15 ppm tert.-butylcatechol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR