4-Methylstyrene 98% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Vinyltoluene, p-Methylstyrene, 4-Vinyltoluene

12770-0025 12770-0100 12770-1000 12770-5000
TS12770-0025EA 193 USD
TS12770-0025 TS12770-0100 TS12770-1000 TS12770-5000
4-Methylstyrene 98% stabilized
4-Methylstyrene
Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 170…175 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0.896 g/cm³
Flash Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008621
CAS Number: 622-97-9
EINECS: 210-762-8
UN: 2618
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5410 to 1.5430 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.1 % TBC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR