3-Nitro-p-toluidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Nitro-4-methylaniline, 4-Methyl-3-nitroaniline, 4-Amino-2-nitrotoluene

12725-0050 12725-0250 12725-1000
TS12725-0050EA 27 USD
TS12725-0050 TS12725-0250 TS12725-1000
3-Nitro-p-toluidine 97%
3-Nitro-p-toluidine
Formula: C₇H₈N₂O₂
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00007910
CAS Number: 119-32-4
EINECS: 204-314-0
UN: 2660
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Ochre to orange to brown Powder
Infrared spectrum Conforms
Melting point 74 to 78 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR