Menadione (2-methyl-1,4-naphthoquinone, vitamin K3) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Menadione, 1,4-Dihydro-2-methylnaphthalene-1,4-dione, 2-Methylnaphthalene-1,4-dione

12718-0050 12718-0250 12718-1000
TS12718-0050EA 25 USD
TS12718-0050 TS12718-0250 TS12718-1000
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3) 98%
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3)
Formula: C₁₁H₈O₂
MW: 172.19 g/mol
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001681
CAS Number: 58-27-5
EINECS: 200-372-6

Order Now

Specification Test Results

Appearance Yellow to green Powder
Infrared spectrum Conforms
Melting point 105 to 108 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR