2-(Methylthio)aniline 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Methylmercapto)aniline

12711-0050 12711-0250
TS12711-0050EA 40 USD
TS12711-0050 TS12711-0250
2-(Methylthio)aniline 97%
2-(Methylthio)aniline
Formula: C₇H₉NS
MW: 139.22 g/mol
Boiling Pt: 234 °C (760 mmHg)
Density: 1.111 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007708
CAS Number: 2987-53-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to brown Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Water ≤0.06 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR