Methyl-2,4-dihydroxybenzoate 97%

Supplier: Thermo Scientific Chemicals
Warning

12673-0050 12673-0250 12673-1000
TS12673-0050EA 31 USD
TS12673-0050 TS12673-0250 TS12673-1000
Methyl-2,4-dihydroxybenzoate 97%
Methyl-2,4-dihydroxybenzoate
Formula: C₈H₈O₄
Melting Pt: 116…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00002276
CAS Number: 2150-47-2
EINECS: 218-428-3

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 116 to 121 °C
GC ≥96.0 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR