4-Methylpyrocatechol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylbenzene-1,2-diol, 4-Methylcatechol

12651-0050 12651-0250 12651-1000 12651-5000
TS12651-0050EA 30 USD
TS12651-0050 TS12651-0250 TS12651-1000 TS12651-5000
4-Methylpyrocatechol 98%
4-Methylpyrocatechol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 251 °C (760 mmHg)
Melting Pt: 62…69 °C
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00002205
CAS Number: 452-86-8
EINECS: 207-214-5

Order Now

Specification Test Results

Appearance White to brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 63 to 68 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR