Isoamyl alcohol 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

12648-0010 12648-0025 12648-0250 12648-5000
TS12648-0010EA 118 USD
TS12648-0010 TS12648-0025 TS12648-0250 TS12648-5000
Isoamyl alcohol 98%, pure
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
Boiling Pt: 130 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.8 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4050 to 1.4070 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR