p-Tolylacetonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

12644-0050 12644-0250 12644-1000
TS12644-0050EA 28 USD
TS12644-0050 TS12644-0250 TS12644-1000
p-Tolylacetonitrile 98%
p-Tolylacetonitrile
Formula: C₉H₉N
Boiling Pt: 242…243 °C (760 mmHg)
Melting Pt: 18 °C
Density: 0.99 g/cm³
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00001922
CAS Number: 2947-61-7
EINECS: 220-963-2
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5170 to 1.5195 (20°C, 589 nm)
GC ≥97 %
Sulfate (SO4) ≤5 ppm
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤10 ppm
Calcium (Ca) ≤5 ppm
Chromium (Cr) ≤0.5 ppm
Iron (Fe) ≤0.5 ppm
Lithium (Li) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤0.5 ppm
Potassium (K) ≤5 ppm
Sodium (Na) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR