4-Methylbenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Aminomethyl)toluene

12640-0050 12640-0250 12640-1000
TS12640-0050EA 32 USD
TS12640-0050 TS12640-0250 TS12640-1000
4-Methylbenzylamine 98%
4-Methylbenzylamine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 13 °C
Density: 0.95 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00008123
CAS Number: 104-84-7
EINECS: 203-243-2
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5330 to 1.5360 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR