Methyl phenylglyoxylate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl benzoylformate, Methyl 2-oxo-2-phenylacetate
12637-0050
12637-0250
TS12637-0050EA
46
USD
TS12637-0050
TS12637-0250
Methyl phenylglyoxylate 99%
Methyl phenylglyoxylate
Formula:
C₉H₈O₃ MW: 164.1600 g/mol Boiling Pt: 246…248 °C (760 mmHg) Melting Pt: 16 °C Density: 1.15 g/cm³ Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008443 CAS Number: 15206-55-0 EINECS: 239-263-3 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5250 to 1.5280 (20°C, 589 nm) |
GC | ≥98.5 % |
Substance darkened by H2SO4 | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...