4-Methylbenzophenone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: p-Methylbenzophenone, Phenyl(p-tolyl)methanone

12635-0500 12635-2500
TS12635-0500EA 63 USD
TS12635-0500 TS12635-2500
4-Methylbenzophenone 97%
4-Methylbenzophenone
Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 53…57 °C
Flash Pt: 143 °C
MDL Number: MFCD00008553
CAS Number: 134-84-9
EINECS: 205-159-1
Merck Index: 12,07470

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR