2-Methylanisole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Cresol methyll ether, 2-Methylanisole, 1-Methoxy-2-methylbenzene, 2-Methoxytoluene

12628-0050 12628-1000 12628-5000
TS12628-0050EA 29 USD
TS12628-0050 TS12628-1000 TS12628-5000
2-Methylanisole 99%
2-Methylanisole
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 172 °C (1013 hPa)
Density: 0.98 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00008373
CAS Number: 578-58-5
EINECS: 209-426-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5180 (20°C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.0005 %
Acidity ≤0.0005 meq/g
Free chlorine ≤0.00003 % (as Cl)
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Aldehyde (HCHO) ≤0.0005 %
E/E ≤0.3 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR