6-Methoxy-1-tetralone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Methoxy-1,2,3,4-tetrahydro-1-naphthalenone, 6-Methoxy-3,4-dihydronaphthalen-1(2H)-one

12612-0250 12612-1000 12612-5000
TS12612-0250EA 57 USD
TS12612-0250 TS12612-1000 TS12612-5000
6-Methoxy-1-tetralone 99%
6-Methoxy-1-tetralone
Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Boiling Pt: 171 °C (11 mmHg)
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00001695
CAS Number: 1078-19-9
EINECS: 214-078-0

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 79 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR