(4-Methoxyphenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

12602-0010 12602-0050 12602-1000 12602-2500
TS12602-0010EA 256 USD
TS12602-0010 TS12602-0050 TS12602-1000 TS12602-2500
(4-Methoxyphenyl)acetic acid 99%
(4-Methoxyphenyl)acetic acid
Formula: (C₆H₄(OCH₃))CH₂COOH
Boiling Pt: 140 °C (3 mmHg)
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00004345
CAS Number: 104-01-8
EINECS: 203-166-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 84 to 88 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR