(3-Methoxyphenyl)acetic acid 99.5%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12601-0050
12601-0250
12601-1000
TS12601-0050EA
32
USD
TS12601-0050
TS12601-0250
TS12601-1000
(3-Methoxyphenyl)acetic acid 99.5%
(3-Methoxyphenyl)acetic acid
Formula:
(C₆H₄(OCH₃))CH₂COOH Melting Pt: 71…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004334 CAS Number: 1798-09-0 EINECS: 217-282-8 |
Specification Test Results
Appearance | White to light beige Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 68 to 72 °C |
Titration with NaOH | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...