2-Nitro-p-anisidine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Amino-3-nitroanisole, 4-Methoxy-2-nitroaniline

12596-0250 12596-1000 12596-5000
TS12596-0250EA 23 USD
TS12596-0250 TS12596-1000 TS12596-5000
2-Nitro-p-anisidine 97%
2-Nitro-p-anisidine
Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00007152
CAS Number: 96-96-8
EINECS: 202-547-2
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 126 °C
HPLC ≥96.0 %
Elemental Analysis 18.13 to 19.07 % N
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR