4-Methoxybenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Aminomethyl)anisole, p-Anisylamine

12578-0250 12578-1000 12578-5000
TS12578-0250EA 63 USD
TS12578-0250 TS12578-1000 TS12578-5000
4-Methoxybenzylamine 98%
4-Methoxybenzylamine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 236…237 °C (760 mmHg)
Density: 1.05 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008122
CAS Number: 2393-23-9
EINECS: 219-247-2
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5460 to 1.5480 (20°C, 589 nm)
GC ≥97.5 %
Sulfide (S) ≤0.0002 %
Carbonate (CO3) ≤0.001 %
Phosphate (PO4) ≤0.00005 %
Sulfate (SO4) ≤0.0002 %
Barium (Ba) ≤0.00001 %
Bismuth (Bi) ≤0.00001 %
Cadmium (Cd) ≤0.000005 %
Calcium (Ca) ≤0.0001 %
Chromium (Cr) ≤0.000005 %
Cobalt (Co) ≤0.00001 %
Copper (Cu) ≤0.00001 %
Gallium (Ga) ≤0.000002 %
Indium (In) ≤0.000002 %
Iron (Fe) ≤0.00001 %
Lead (Pb) ≤0.000005 %
Lithium (Li) ≤0.000002 %
Magnesium (Mg) ≤0.00005 %
Manganese (Mn) ≤0.00001 %
Molybdenum (Mo) ≤0.00001 %
Nickel (Ni) ≤0.000005 %
Platinum (Pt) ≤0.00001 %
Potassium (K) ≤0.0001 %
Sodium (Na) ≤0.0001 %
Strontium (Sr) ≤0.0001 %
Thallium (Tl) ≤0.000005 %
Tin (Sn) ≤0.00001 %
Titanium (Ti) ≤0.00001 %
Zinc (Zn) ≤0.00001 %
Reducing substances ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR