2-Methoxybenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Hydroxymethyl)anisole, (2-Methoxyphenyl)methanol

12576-0100 12576-0500 12576-2500
TS12576-0100EA 27 USD
TS12576-0100 TS12576-0500 TS12576-2500
2-Methoxybenzyl alcohol 99%
2-Methoxybenzyl alcohol
Formula: C₈H₁₀O₂
Boiling Pt: 248…250 °C (760 mmHg)
Density: 1.03 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004611
CAS Number: 612-16-8
EINECS: 210-296-5

Order Now

Specification Test Results

Appearance Clear colorless to Yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5460 to 1.5490 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR