4-Methoxybenzophenone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: p-Methoxybenzophenone, (4-Methoxyphenyl)(phenyl)methanone

12574-0250 12574-1000
TS12574-0250EA 60 USD
TS12574-0250 TS12574-1000
4-Methoxybenzophenone 97%
4-Methoxybenzophenone
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 58…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00008403
CAS Number: 611-94-9
EINECS: 210-285-5

Order Now

Specification Test Results

Appearance White to yellow to light orange Crystalline solid
Infrared spectrum Conforms
Melting point 58 to 63 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR