2,4,6-Trimethylbenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12557-0100
12557-0500
TS12557-0100EA
87
USD
TS12557-0100
TS12557-0500
2,4,6-Trimethylbenzaldehyde 97%
2,4,6-Trimethylbenzaldehyde
Formula:
C₁₀H₁₂O Boiling Pt: 237 °C (1013 hPa) Melting Pt: 14 °C Density: 1 g/cm³ Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003341 CAS Number: 487-68-3 EINECS: 207-662-1 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5520 to 1.5540 (20°C, 589 nm) |
GC | ≥96.0 % |
Loss on ignition | 24.5 % to 26.5 % (800°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...