4-Iodoanisole 98%

Supplier: Thermo Scientific Chemicals

12232-0250 12232-1000 12232-5000
TS12232-0250EA 41 USD
TS12232-0250 TS12232-1000 TS12232-5000
4-Iodoanisole 98%
4-Iodoanisole
Formula: C₇H₇IO
Boiling Pt: 237 °C (726 mmHg)
Melting Pt: 46…51 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001056
CAS Number: 696-62-8
EINECS: 211-798-7

Order Now

Specification Test Results

Appearance Off-white to brown Crystalline powder and/or chunks (May darken on storage)
Infrared spectrum Conforms
Melting point 46 to 51 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR